missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IFT57 Monoclonal Antibody (OTI5B6), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA500906S
This item is not returnable.
View return policy
Description
IFT57 Monoclonal antibody specifically detects IFT57 in Human samples. It is validated for Immunohistochemistry (Paraffin), Western BlotSpecifications
| IFT57 | |
| Monoclonal | |
| 0.53 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q9NZ45 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 55847 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG1 |
| Immunohistochemistry (Paraffin), Western Blot | |
| OTI5B6 | |
| Unconjugated | |
| cisd1 | |
| 4833420A15Rik; dermal papilla-derived protein 8; DERP8; Esrrbl1; estrogen-related receptor beta like 1; estrogen-related receptor beta-like protein 1; FLJ10147; HIP1 protein interactor; HIP1-interacting protein; Hippi; huntingtin interacting protein-1 interacting protein; huntingtin-interacting protein-1 protein interactor; IFT57; intraflagellar transport 57; intraflagellar transport 57 homolog; intraflagellar transport protein 57 homolog; MHS4R2; Vestrogen-related receptor beta like 1 | |
| Full length human recombinant protein of human CISD1 produced in HEK293T cell. | |
| 30 μL | |
| Primary | |
| Human | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction