missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IFT74 Antibody, Novus Biologicals™
Goat Polyclonal Antibody has been used in 2 publications
Brand: Novus Biologicals NB300-822
This item is not returnable.
View return policy
Description
IFT74 Polyclonal specifically detects IFT74 in Human samples. It is validated for Western Blot, Peptide ELISA.
Specifications
| IFT74 | |
| Polyclonal | |
| Unconjugated | |
| Tris saline (20 mM Tris pH 7.3, 150 mM NaCl), 0.5% BSA with 0.02% Sodium Azide | |
| Capillary morphogenesis gene 1 protein, CCDC2capillary morphogenesis protein 1, CMG1Coiled-coil domain-containing protein 2, CMG-1MGC111562, coiled-coil domain containing 2, FLJ22621, intraflagellar transport 74 homolog (Chlamydomonas), intraflagellar transport protein 74 homolog | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 80173 | |
| Store at -20C. Avoid freeze-thaw cycles. |
| Western Blot, ELISA, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| 0.5 mg/mL | |
| Western Blot 0.2 - 0.6 ug/mL, Peptide ELISA Detection limit 1:32000 | |
| NP_079379 | |
| IFT74 | |
| Peptide with sequence C-KTIVDALHSTSGN corresponding to C-Terminus according to NP_001092692.1, NP_001092693.1, NP_079379.2, AAK77221.1. | |
| 0.1 mg | |
| Primary | |
| Human, Rat | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction