missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ IFT81 Polyclonal Antibody
GREENER_CHOICE

Product Code. 17160044 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17160044 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17160044 Supplier Invitrogen™ Supplier No. PA5112184

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

IFT81 Polyclonal Antibody for Western Blot, IHC (P), IP

Isoform CDV-1 may be involved in cardiac hypertrophy caused by carnitine deficiency. Isoform CDV-1R appears to play a role in development of the testis and spermatogenesis.
TRUSTED_SUSTAINABILITY

Specifications

Antigen IFT81
Applications Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.03% ProClin 300; pH 7
Gene IFT81
Gene Accession No. Q8WYA0
Gene Alias AW060663; carnitine deficiency-associated gene expressed in ventricle 1; carnitine deficiency-associated protein expressed in ventricle 1; Cdv1; CDV-1; CDV1R; CDV-1R; DV1; Ift81; intraflagellar transport 81; intraflagellar transport 81 homolog; intraflagellar transport protein 81 homolog
Gene Symbols IFT81
Host Species Rabbit
Immunogen E. coli-derived Human IFT81 fragment.
Purification Method Antigen Affinity Chromatography, Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 28981
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.