Learn More
IFT88 Polyclonal Antibody
GREENER_CHOICE
IFT88 Polyclonal Antibody
Brand:  Invitrogen PA599962
Product Code. 16684205
  • € 502.00 / 100µL
Description
IFT88 Polyclonal Antibody for Western Blot, IHC (P)
TRUSTED_SUSTAINABILITY
Specifications
Specifications
IFT88
Polyclonal
Unconjugated
IFT88
AW552028; D13S1056E; DAF19; fb37b11; flexo; fxo; HGNC:20606; hTg737; Ift88; intraflagellar transport 88; intraflagellar transport 88 homolog; intraflagellar transport protein; intraflagellar transport protein 88; intraflagellar transport protein 88 homolog; MGC26259; orpk; osm-5; oval; ovl; polaris; polaris homolog; polaris; axonemal transport protein; probe hTg737 (polycystic kidney disease, autosomal recessive); recessive polycystic kidney disease protein Tg737; Recessive polycystic kidney disease protein Tg737 homolog; RP11-172H24.2; testicular tissue protein Li 93; tetratricopeptide repeat domain 10; tetratricopeptide repeat protein 10; TG737; Tg737 protein; Tg737Rpw; TgN(Imorpk)737Rpw; TgN737Rpw; TPR repeat protein 10; transgene insert site 737, insertional mutation, polycystic kidney disease; Ttc10; wu:fb37b11; xx:zah8127
Rabbit
Affinity Chromatography
RUO
21821, 305918, 477345, 8100
-20°C
Liquid
Immunohistochemistry (Paraffin), Western Blot
1 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Q13099, Q61371
IFT88
A synthesized peptide derived from human IFT88, corresponding to a region within C-terminal amino acids.
100 μL
Primary
Human, Mouse, Rat, Canine