missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ig gamma-4 Chain C Region Antibody (rIGHG4/1345), DyLight 650, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-08668C
This item is not returnable.
View return policy
Description
Ig gamma-4 Chain C Region Monoclonal antibody specifically detects Ig gamma-4 Chain C Region in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
| Ig gamma-4 Chain C Region | |
| Monoclonal | |
| DyLight 650 | |
| IGHG4, mmunoglobulin heavy constant gamma 4, mmunoglobulin heavy constant gamma 4 (G4m marker) | |
| Recombinant human Ig gamma-4 Chain C Region fragment (Uniprot: P01861) | |
| 100 μg | |
| Primary | |
| Human | |
| Purified |
| Immunohistochemistry (Paraffin) | |
| rIGHG4/1345 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| 3503 | |
| Store at 4°C in the dark. | |
| IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction