missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IgA Antibody (RM220) - Azide and BSA Free, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-25955
This item is not returnable.
View return policy
Description
IgA Monoclonal antibody specifically detects IgA in Mouse samples. It is validated for Western Blot, ELISA
Specifications
| IgA | |
| Monoclonal | |
| Unconjugated | |
| PBS | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| Mouse | |
| Purified |
| Western Blot, ELISA | |
| RM220 | |
| Western Blot : 0.5 ug/ml-2 ug/ml, ELISA : 0.005 ug/ml - 0.2 ug/ml | |
| Immunoglobulin A | |
| Mouse IgA | |
| 100 μg | |
| Adaptive Immunity, Immunology | |
| 3493 | |
| Store at -20C. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction