missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IgA Antibody (RM220), Biotin, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP2-62011B
This item is not returnable.
View return policy
Description
IgA Monoclonal antibody specifically detects IgA in Mouse samples. It is validated for Western Blot, ELISA
Specifications
| IgA | |
| Monoclonal | |
| 1 mg/mL | |
| Western Blot 0.5 μg/mL - 2 μg/mL, ELISA 0.005 μg/mL - 0.2 μg/mL | |
| Immunoglobulin A | |
| Mouse IgA | |
| 50 μg | |
| Adaptive Immunity, Immunology | |
| 3493 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot, ELISA | |
| RM220 | |
| Biotin | |
| PBS, 50% Glycerol, 1% BSA | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction