Learn More
IgA Goat anti-Dog, Polyclonal , Abnova™
Goat polyclonal antibody raised against dog IgA.
Marque:  Abnova PAB29763.1mg
Code produit. 16048207
  • € 188.00 / 1mg
Les retours ne sont pas autorisés pour ce produit. Afficher la politique du retour.
Description
Description
Spécification
Spécification
IgA
Polyclonal
Goat polyclonal antibody raised against dog IgA.
In PBS, pH 7.2 (0.09% sodium azide)
Dog IgA whole molecule.
1 mg
Secondary
Dog
Liquid
ELISA, Immunoprecipitation, Western Blot
Unconjugated
ELISA (2 ug/mL) Immunoprecipitation (2 ug/mg) Western Blot (0.05-0.5 ug/mL) The optimal working dilution should be determined by the end user.
Goat
Antigen Affinity Purified
RUO
The affinity purified antibodies react only with IgA by immunodiffusion and IEP techniques.
Store at 4°C.