missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IgA Heavy Chain Antibody (IGHA/7567R), Novus Biologicals™
Rabbit Monoclonal Antibody
€ 286.00 - € 530.00
Specifications
| Antigen | IgA Heavy Chain |
|---|---|
| Clone | IGHA/7567R |
| Concentration | 0.2 mg/ml |
| Dilution | Immunohistochemistry-Paraffin 1-2 μg/mL |
| Applications | Immunohistochemistry (Paraffin) |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18620925
|
Novus Biologicals
NBP3-20648-100ug |
100 μg |
€ 530.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18675244
|
Novus Biologicals
NBP3-20648-20ug |
20 μg |
€ 286.00
20µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
IgA Heavy Chain Monoclonal antibody specifically detects IgA Heavy Chain in Human samples. It is validated for Immunohistochemistry (Paraffin)Specifications
| IgA Heavy Chain | |
| 0.2 mg/ml | |
| Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| Human | |
| Full-length native protein (purified) corresponding to Human IgA | |
| Primary | |
| Store at 4&drg;C. Do not freeze. |
| IGHA/7567R | |
| Immunohistochemistry-Paraffin 1-2 μg/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| 10mM PBS with 0.05% BSA | |
| IgG κ | |
| Protein A or G purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title