missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IgA (Heavy Chain) Mouse anti-Human, Clone: SPM187, Abnova™
Mouse monoclonal antibody raised against native human IgA (Heavy Chain).
Brand: Abnova MAB13094.100ug
This item is not returnable.
View return policy
Specifications
| IgA (Heavy Chain) | |
| Monoclonal | |
| Unconjugated | |
| Flow Cytometry (1-2 ug/106 cells in 0.1 mL) Immunofluorescence (0.5-1 ug/mL) Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (0.5-1 ug/mL) The optimal working dilution should be determined by the end user. | |
| FLJ14473/FLJ35065/FLJ35500/FLJ36402/FLJ39698/FLJ40001/FLJ41548/FLJ41552/FLJ41789/FLJ43248/FLJ43594/FLJ44293/FLJ46028/FLJ46621/FLJ46724/FLJ46811/FLJ46824/IgA1/MGC102857 | |
| Mouse | |
| Protein A/G purification | |
| RUO | |
| 3493/3494 | |
| Human | |
| Liquid |
| Flow Cytometry, Immunofluorescence, Immunohistochemistry (PFA fixed) | |
| SPM187 | |
| Mouse monoclonal antibody raised against native human IgA (Heavy Chain). | |
| In 10mM PBS. | |
| IGHA1 | |
| Native purified human IgA. | |
| 100 μg | |
| Primary | |
| This monoclonal antibody is specific to heavy chain of IgA and shows minimal cross-reaction with heavy chains of other immunoglobulins. | |
| Store at -20 to -80°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction