Learn More
IGBP1 Antibody, DyLight 680, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand:  Novus Biologicals NB100-487FR
Product Code. 18753023
  • € 633.00 / 0.10mL
Description
IGBP1 Polyclonal antibody specifically detects IGBP1 in Human, Mouse samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY
Specifications
Specifications
The immunogen recognized by this antibody maps to a region between residue 300 and the C-terminus (residue 339) of human Protein Phosphatase 2A, regulatory subunit alpha-4 (Immunoglobulin binding protein 1) using the numbering given in SwissProt entry P78318 (GeneID 3476).
3476