missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IgD Antibody (11-26C), Janelia Fluor™ 646, Novus Biologicals™
Rat Monoclonal Antibody
Brand: Novus Biologicals FAB9858J
This item is not returnable.
View return policy
Description
IgD Monoclonal antibody specifically detects IgD in Mouse samples. It is validated for Flow Cytometry, CyTOF
Specifications
| IgD | |
| Monoclonal | |
| Janelia Fluor 646 | |
| 50mM Sodium Borate | |
| Rat | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| Primary | |
| Mouse | |
| Purified |
| Flow Cytometry, CyTOF | |
| 11-26C | |
| Flow Cytometry, CyTOF-ready | |
| IgD, Igh-5, Ighd, Immunoglobulin D, Immunoglobulin Delta | |
| Mouse IgD | |
| 0.1 mL | |
| B Cell Development and Differentiation Markers, Immunology | |
| 3495 | |
| Store at 4C in the dark. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction