Learn More
IgD Antibody (IA6-2) [Janelia Fluor™ 646], Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals FAB9857J
Product Code. 18556779
  • ONLINE EXCLUSIVE PRICE
    € 663.00
    € 626.85 / 0.10mL
    Save € 36.15
    5% Off
Description
IgD Monoclonal antibody specifically detects IgD in Human samples. It is validated for Flow Cytometry, CyTOF
TRUSTED_SUSTAINABILITY
Specifications
Specifications
IgD
Monoclonal
Janelia Fluor 646
50mM Sodium Borate
Mouse
Protein A or G purified from hybridoma culture supernatant
RUO
Primary
Human
Purified
Flow Cytometry, CyTOF
IA6-2
Flow Cytometry, CyTOF-ready
IgD, Igh-5, Ighd, Immunoglobulin D, Immunoglobulin Delta
Human IgD,
0.1 mL
B Cell Development and Differentiation Markers, Immunology
3495
Store at 4°C in the dark.
IgG2a