missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IgD Antibody (IgD26), Alexa Fluor™ 700, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-50437AF700
This item is not returnable.
View return policy
Description
IgD Monoclonal specifically detects IgD in Human samples. It is validated for Flow Cytometry, Immunohistochemistry.
Specifications
| IgD | |
| Monoclonal | |
| Alexa Fluor 700 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| IGHD | |
| IgD purified from human myeloma serum | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
| Flow Cytometry, Immunohistochemistry | |
| IgD26 | |
| Flow Cytometry, Immunohistochemistry | |
| IgD, Igh-5, Ighd, Immunoglobulin D, Immunoglobulin Delta | |
| Mouse | |
| Protein A purified | |
| RUO | |
| 3495 | |
| Store at 4C in the dark. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction