missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IgD Antibody (IGHD/6818R), PE, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-20992PE
This item is not returnable.
View return policy
Description
IgD Monoclonal antibody specifically detects IgD in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
| IgD | |
| Monoclonal | |
| PE | |
| IgD, Igh-5, Ighd, Immunoglobulin D, Immunoglobulin Delta | |
| Recombinant fragment (around aa1-200) of human IgD protein (exact sequence is proprietary) | |
| 0.1 mL | |
| B Cell Development and Differentiation Markers, Immunology | |
| 3495 | |
| Store at 4°C in the dark. | |
| IgG κ |
| Immunohistochemistry (Paraffin) | |
| IGHD/6818R | |
| PBS | |
| Rabbit | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction