missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IGDCC3 Antibody (920038) [Janelia Fluor™ 669], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals FAB8559JF669
This item is not returnable.
View return policy
Description
IGDCC3 Monoclonal antibody specifically detects IGDCC3 in Human samples. It is validated for Flow Cytometry,Western Blot,Immunocytochemistry,CyTOF
Specifications
| IGDCC3 | |
| Monoclonal | |
| Janelia Fluor 669 | |
| immunoglobulin superfamily DCC subclass member 3, immunoglobulin superfamily, DCC subclass, member 3, Putative neuronal cell adhesion moleculePUNCHsT18880 | |
| Human embryonic kidney cell line HEK293-derived recombinant human IGDCC3, Met1-Gly640, Accession # Q81VU1 | |
| RUO | |
| 9543 | |
| Store at 4°C in the dark. | |
| IgG1 |
| Flow Cytometry, Western Blot, Immunocytochemistry, CyTOF | |
| 920038 | |
| 50mM Sodium Borate | |
| Mouse | |
| 0.1 mL | |
| Primary | |
| Human | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction