missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IgE Antibody (5D4cc) [Janelia Fluor™ 585], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NB110-8331JF585
This item is not returnable.
View return policy
Description
IgE Monoclonal antibody specifically detects IgE in Human samples. It is validated for ELISA,Sandwich ELISA
Specifications
| IgE | |
| Monoclonal | |
| Janelia Fluor 585 | |
| IgE, immunoglobulin heavy constant epsilon | |
| Hybridoma clone has been derived from hybridization of Sp2/0 myeloma cells with spleen cells of Balb/c mice immunized with IgE from myelomatous human serum. | |
| 0.2 mL | |
| Adaptive Immunity, Immunology | |
| 3497 | |
| Store at 4°C in the dark. | |
| IgG2a |
| ELISA, Sandwich ELISA | |
| 5D4cc | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction