Learn More
IgE Mouse anti-Human, Brilliant Violet 510, Clone: G7-26, BD Optibuild™
Mouse Monoclonal Antibody
Brand: BD Biosciences 744316
Description
The G7-26 monoclonal antibody specifically recognizes the constant region of human Immunoglobulin E (IgE). It does not crossreact with other immunoglobulin heavy chain isotypes. IgE exists in a transmembrane form that is expressed by B lymphocytes and serves as an antigen receptor. Soluble IgE is produced and secreted by activated B cells and plasma cells. IgE may bind through its constant region to cell surface receptors, such as the high-affinity ( ) or low-affinity ( /CD23) receptors for IgE. Fc RI is expressed on mast cells, basophils, and at lower levels, on dendritic cells and monocytes.
The antibody was conjugated to BD Horizon™ BV510 which is part of the BD Horizon Brilliant™ Violet family of dyes. With an Ex Max of 405nm and Em Max at 510nm, BD Horizon BV510 can be excited by the violet laser and detected in the BD Horizon V500 (525/50nm) filter set. BD Horizon BV510 conjugates are useful for the detection of dim markers off the violet laser. BV510 is a polymer-based dye that is brighter than BD Horizon V500. Due to similar excitation and emission properties, BD Horizon BV510 and BD Horizon V500 cannot be used simultaneously.
Specifications
| IgE | |
| G7-26 | |
| Brilliant Violet 510 | |
| Immunoglobulin E, IGHE, IGHε, Ig epsilon chain constant region | |
| Affinity Purified | |
| RUO | |
| Human | |
| IgG2a κ |
| Monoclonal | |
| 0.2mg/mL | |
| Aqueous buffered solution containing ≤0.09% sodium azide. | |
| Mouse | |
| 50 μg | |
| Primary | |
| Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.