missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LAS2 Antibody (OTI2B2), Alexa Fluor™ 488, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-72452AF488
This item is not returnable.
View return policy
Description
LAS2 Monoclonal specifically detects LAS2 in Human samples. It is validated for Western Blot.
Specifications
| LAS2 | |
| Monoclonal | |
| Alexa Fluor 488 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| C18orf54 | |
| Full length recombinant protein of human LAS2 (NP_775800) produced in E.coli. | |
| 0.1 mL | |
| 162681.0 | |
| Store at 4°C in the dark. |
| Western Blot | |
| OTI2B2 | |
| Western Blot | |
| C18orf54, chromosome 18 open reading frame 54, hypothetical protein LOC162681, LAS2, MGC33382 | |
| Mouse | |
| Affinity Purified | |
| Primary | |
| Human | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction