missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ IGF-I R/IGF1R Antibody (dalotuzumab) - Humanized, Novus Biologicals™
Human Monoclonal Antibody
Brand: Novus Biologicals™ NBP3-28286-1mg
This item is not returnable.
View return policy
Description
IGF-I R/IGF1R Monoclonal antibody specifically detects IGF-I R/IGF1R in Human samples. It is validated for Flow Cytometry,ELISA,Functional Assay
Specifications
| IGF-I R/IGF1R | |
| Monoclonal | |
| Unconjugated | |
| P08069 | |
| Human | |
| Protein A purified | |
| RUO | |
| Primary | |
| Reconstitute with sterile, distilled water to a final concentration of 1 mg/mL. Gently shake to solubilize completely. Do not vortex. | |
| Store at -20°C in powder form. Store at -80°C once reconstituted. | |
| IgG1 |
| Flow Cytometry, ELISA, Functional Assay | |
| dalotuzumab | |
| Lyophilized from 25mM histidine, 8% sucrose, 0.01% Tween80 (pH6.2) | |
| CD221, CD221 antigen, EC 2.7.10, EC 2.7.10.1, IGF-I receptor, IGFIR, IGFR, insulin-like growth factor 1 receptor, Insulin-like growth factor I receptor, JTK13, MGC142170, MGC142172, MGC18216, soluble IGF1R variant 1, soluble IGF1R variant 2 | |
| IGF1R/ CD221 | |
| 1 mg | |
| Apoptosis, Cancer, Lipid and Metabolism, Neuroscience, Phospho Specific, Protein Kinase, Signal Transduction, Tyrosine Kinases | |
| 3480 | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction