Learn More
IGF-I R/IGF1R Antibody (Immunomedics patent anti-IGF-1R), CoraFluor™ 1, Novus Biologicals™
Human Monoclonal Antibody
Brand:  Novus Biologicals NBP3-28494CL1
Product Code. 30154381
  • € 869.00 / 0.10mL
Description
IGF-I R/IGF1R Monoclonal antibody specifically detects IGF-I R/IGF1R in Human samples. It is validated for Flow Cytometry,ELISA,Functional Assay
TRUSTED_SUSTAINABILITY
Specifications
Specifications
IGF-I R/IGF1R
Monoclonal
CoraFluor 1
CD221, CD221 antigen, EC 2.7.10, EC 2.7.10.1, IGF-I receptor, IGFIR, IGFR, insulin-like growth factor 1 receptor, Insulin-like growth factor I receptor, JTK13, MGC142170, MGC142172, MGC18216, soluble IGF1R variant 1, soluble IGF1R variant 2
IGF1R/ CD221
0.1 mL
Apoptosis, Cancer, Lipid and Metabolism, Neuroscience, Phospho Specific, Protein Kinase, Signal Transduction, Tyrosine Kinases
3480
Store at 4°C in the dark. Do not freeze.
IgG1
Flow Cytometry, ELISA, Functional Assay
Immunomedics patent anti-IGF-1R
PBS
Human
Protein A purified