Learn More
PLA2G3 Monoclonal Antibody (OTI4F2), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand:  Origene Technologies TA811652
Encompass_Preferred
 View more versions of this product
Product Code. 17595794
  • € 540.87 / 100µL
Description
PLA2G3 belongs to the family of secreted phospholipase A2 (sPLA2; EC 3.1.1.4) proteins. These Ca(2+)-dependent lipolytic enzymes have a conserved Ca(2+)-binding loop and a his-asp dyad in the catalytic site.
Specifications
Specifications
PLA2G3
Monoclonal
1 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Q9NZ20
PLA2G3
Human recombit protein fragment corresponding to amino acids 409-509 of human PLA2G3 produced in E.coli.
100 μL
Primary
Human
Antibody
IgG1
Western Blot
OTI4F2
Unconjugated
PLA2G3
GIII-SPLA2, SPLA2III, sPLA2-III
Mouse
Affinity Chromatography
RUO
50487
-20° C, Avoid Freeze/Thaw Cycles