Learn More
Invitrogen™ IGF2-AS Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5144025
Description
Adding 0.2 mL of distilled water will yield a concentration of 500 μg/mL. Positive Control - WB: human PANC-1 whole cell. ICC/IF: U20S cell. Flow: CACO-2 cell. Store at -20°C for one year from date of receipt. After reconstitution, at 4°C for one month. It can also be aliquotted and stored frozen at -20°C for six months. Avoid repeated freeze-thaw cycles.
The insulin-like growth factors possess growth-promoting activity. In vitro, they are potent mitogens for cultured cells. IGF-II is influenced by placental lactogen and may play a role in fetal development. Preptin undergoes glucose-mediated co-secretion with insulin, and acts as physiological amplifier of glucose-mediated insulin secretion. Exhibits osteogenic properties by increasing osteoblast mitogenic activity through phosphoactivation of MAPK1 and MAPK3.
Specifications
| IGF2-AS | |
| Polyclonal | |
| Unconjugated | |
| IGF2-AS | |
| IGF; IGF2 antisense RNA; IGF2AS; IGF2-AS; IGF2-AS1; Insulin like growth factor; PEG8; PEG8/IGF2AS protein | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 51214 | |
| -20°C | |
| Lyophilized |
| ELISA, Flow Cytometry, Western Blot, Immunocytochemistry | |
| 500 μg/mL | |
| PBS with 4mg trehalose and 0.01mg sodium azide | |
| Q6U949 | |
| IGF2-AS | |
| E.coli-derived human IGF2-AS recombinant protein (Position: M1-K168). | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.