missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IGF2BP2 Monoclonal Antibody (OTI6C4), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA501273
This item is not returnable.
View return policy
Description
IGF2BP2 is a member of the IGF-II mRNA-binding protein (IMP) family. It recruits target transcripts to cytoplasmic protein-RNA complexes. The protein contains several four KH domains and two RRM domains. It functions by binding to the 5′ UTR of the insulin-like growth factor 2 (IGF2) mRNA and regulating IGF2 translation. Alternate transcriptional splice variants, encoding different isoforms, have been characterized. Autoantibodies against IGF2BP2 are detected in sera from some patients with hepatocellular carcinoma.Specifications
| IGF2BP2 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q9Y6M1 | |
| IGF2BP2 | |
| Full length human recombit protein of human IGF2BP2 produced in HEK293T cell. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Immunocytochemistry, Immunofluorescence, Western Blot | |
| OTI6C4 | |
| Unconjugated | |
| IGF2BP2 | |
| IMP-2, IMP2, VICKZ2 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 10644 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction