missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IGF2BP3 Antibody (8F11), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP1-51674
This item is not returnable.
View return policy
Description
IGF2BP3 Monoclonal specifically detects IGF2BP3 in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| IGF2BP3 | |
| Monoclonal | |
| Unconjugated | |
| CT98, hKOC, IGF II mRNA binding protein 3, IGF2 mRNA-binding protein 3, IGF-II mRNA-binding protein 3, IMP-3KH domain-containing protein overexpressed in cancer, IMP3VICKZ family member 3, insulin-like growth factor 2 mRNA binding protein 3, insulin-like growth factor 2 mRNA-binding protein 3, KH domain containing protein overexpressed in cancer, KOC1DKFZp686F1078, VICKZ3cancer/testis antigen 98 | |
| Mouse | |
| 69 kDa | |
| 0.1 mL | |
| Cancer | |
| 10643 | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| IgG1 |
| Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| 8F11 | |
| Western Blot 1:500-1:2000, ELISA 1:10000, Immunohistochemistry 1:200-1:1000, Immunohistochemistry-Paraffin 1:200-1:1000 | |
| IGF2BP3 | |
| Purified recombinant fragment of human IGF2BP3 expressed in E. Coli. | |
| Unpurified | |
| RUO | |
| Primary | |
| Human | |
| Ascites |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction