missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IGF2BP3 Mouse anti-Human, Clone: OTI4H5, liquid, TrueMAB™
Mouse Monoclonal Antibody
Brand: Origene Technologies TA811487
This item is not returnable.
View return policy
Description
Insulin-like growth factor-II messenger RNA (mRNA)-binding protein-3 (IMP-3), also known as K homology domain-containing protein overexpressed in cancer (KOC) and L523S, is a member of the insulin-like growth factor-II mRNA-binding protein family and is expressed during embryogenesis and in some maligcies. IMP-3 is expressed in maligt melanoma but not in benign nevi, even when dysplastic features are present; IMP-3 is expressed in a significantly higher proportion of melanomas than Spitz nevi; and IMP-3 is expressed in metastatic melanomas significantly more than in thin melanomas. IMP-3 appears to be involved in the progression of maligt melanoma and may play an important role in the regulation of the biologic behavior of this tumor.Specifications
| IGF2BP3 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| O00425 | |
| IGF2BP3 | |
| Full length human recombit protein of human IGF2BP3 produced in E.coli. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Immunohistochemistry (Paraffin), Western Blot | |
| OTI4H5 | |
| Unconjugated | |
| IGF2BP3 | |
| CT98, IMP-3, IMP3, KOC, KOC1, VICKZ3 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 10643 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction