missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IGF2R Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 20253-1-AP-20UL
This item is not returnable.
View return policy
Description
The antibody is specific to IGF2R.
Lysosomal enzymes containing one or two mannose 6-phosphate (man6P) moieties are moved about in the cell by two distinct but interconnected cycles by means of approximately 300 kDa cation independent mannose 6-phospate receptors (CI-MPR). The biosynthetic cycle refers to the MPR transport of newly synthesized lysosomal enzymes from the trans Golgi network to late endosomes or early lysosomes. The endocytic cycle transports extracellular lysosomal enzymes from the plasma membrane via clathrin-coated vesicles to early endosomes. The entire pool of MPRs cycles between these cellular compartments every 3 hours. The steady state distribution of MPR′s is predomitly within late endosomes, fewer in the trans Golgi network and approximately 10 % at cell surface. In addition to its man6P binding activity, the MPR contains a separate binding site for the type II insulin-like growth factor and is capable of binding both man6P and IGF-II simultaneously. An approximately 240 kDa soluble, truncated form, representing the extracellular domain of the protein, has also been found circulating in serum and is capable of binding both ligands.Specifications
| IGF2R | |
| Polyclonal | |
| Unconjugated | |
| IGF2R | |
| CD222, CI Man 6 P receptor, CI MPR, CIMPR, IGF II receptor, IGF2R, M6P R, M6P/IGF2 receptor, M6P/IGF2R, M6PR, MPR 300, MPR1, MPRI | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 3482 | |
| -20°C | |
| Liquid |
| Flow Cytometry, Immunoprecipitation, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| 0.42 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P11717 | |
| IGF2R | |
| Peptide | |
| 20 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction