Learn More
IGF2R-Specific Rabbit anti-Human, CoraLite 488, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech CL488-20253-100UL
Product Code. 17402523
  • € 450.00 / 100µL
Description
The IGF2R-Specific antibody from Proteintech is a rabbit polyclonal antibody generated with peptide of human IGF2R-Specific. This antibody recognizes human antigen. The IGF2R-Specific antibody has been validated for the following applications: FC (Intra) analysis. This antibody is coralite™488.
Specifications
Specifications
IGF2R-Specific
Polyclonal
Rabbit
Primary
Human
Flow Cytometry (Intracellular Staining)
CoraLite 488
100 μL
3482