missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IGFBP-2 Antibody (7G11), Novus Biologicals™
Rabbit Monoclonal Antibody
€ 343.00 - € 533.00
Specifications
| Antigen | IGFBP-2 |
|---|---|
| Clone | 7G11 |
| Dilution | Western Blot 1:500-1:5000, ELISA, Immunohistochemistry 1:50-1:200 |
| Applications | Western Blot, ELISA, Immunohistochemistry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30032191
|
Novus Biologicals
NBP3-26207-50ul |
50 μL |
€ 343.00
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30028636
|
Novus Biologicals
NBP3-26207-100ul |
100 μL |
€ 533.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
IGFBP-2 Monoclonal antibody specifically detects IGFBP-2 in Human samples. It is validated for Western Blot, ELISA, ImmunohistochemistrySpecifications
| IGFBP-2 | |
| Western Blot 1:500-1:5000, ELISA, Immunohistochemistry 1:50-1:200 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| BP2, IBP2, IBP-2, IGF-binding protein 2, IGFBP-2, IGF-BP53, insulin-like growth factor binding protein 2 (36kD), insulin-like growth factor binding protein 2, 36kDa, insulin-like growth factor-binding protein 2 | |
| A synthesized peptide derived from Human IGFBP-2 [UniProt P18065] | |
| Primary | |
| Store at -20 to -70C. Avoid freeze-thaw cycles. |
| 7G11 | |
| Western Blot, ELISA, Immunohistochemistry | |
| Unconjugated | |
| Rabbit | |
| Cancer, Cell Biology, Cytokines and Growth Factors, Signal Transduction | |
| PBS, pH 7.4, 150mM NaCl and 50% glycerol | |
| 3485 | |
| IgG | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title