missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IGFBP-3 Antibody (008), Allophycocyanin, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP2-89495APC
This item is not returnable.
View return policy
Description
IGFBP-3 Monoclonal antibody specifically detects IGFBP-3 in Human samples. It is validated for ELISA, ELISA
Specifications
| IGFBP-3 | |
| Monoclonal | |
| APC | |
| PBS | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| ELISA, Sandwich ELISA | |
| 008 | |
| ELISA, Sandwich ELISA Detection | |
| acid stable subunit of the 140 K IGF complex, binding protein 29, binding protein 53, growth hormone-dependent binding protein, IBP-3, IBP3BP-53, IGF-binding protein 3, IGFBP-3, insulin-like growth factor binding protein 3, insulin-like growth factor-binding protein 3 | |
| This antibody was obtained from a rabbit immunized with purified, recombinant Human IGFBP-3 (Gly28-Lys 291; Accession#: NP_000589.2). | |
| 0.1 mL | |
| Apoptosis, Biologically Active Proteins, Prostate Cancer | |
| 3486 | |
| Store at 4C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction