Learn More
IGFBP-6 Antibody (001), Alexa Fluor™ 647, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand:  Novus Biologicals NBP2-90157AF647
Product Code. 18228778
  • € 617.00 / 0.10mL
Description
IGFBP-6 Monoclonal specifically detects IGFBP-6 in Human samples. It is validated for ELISA, Sandwich ELISA Detection.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
IGFBP-6
Monoclonal
Alexa Fluor 647
50mM Sodium Borate
Rabbit
Protein A purified
RUO
Primary
Human
Purified
ELISA
1
ELISA, Sandwich ELISA Detection
IBP6, IBP-6, IGF binding protein 6, IGF-binding protein 6, IGFBP-6, insulin-like growth factor binding protein 6, insulin-like growth factor-binding protein 6
This antibody was obtained from a rabbit immunized with purified, recombinant Human IGFBP-6 (P24592; Met1-Gly240).
0.1 mL
Cell Cycle and Replication
3489
Store at 4°C in the dark.