missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IGFBP1 Monoclonal Antibody (OTI6C7), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA808733S
This item is not returnable.
View return policy
Description
IGFBP1 Monoclonal antibody specifically detects IGFBP1 in Human samples. It is validated for Immunohistochemistry (Paraffin), Western BlotSpecifications
| IGFBP1 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q6FI81 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 57019 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG1 |
| Immunohistochemistry (Paraffin), Western Blot | |
| OTI6C7 | |
| Unconjugated | |
| CIAPIN1 | |
| AFBP; alpha-pregnancy-associated endometrial globulin; amniotic fluid binding protein; Binding protein 25; Binding protein 26; Binding protein 28; binding protein-25; binding protein-26; binding protein-28; growth hormone independent-binding protein; hIGFBP 1; hIGFBP1; hIGFBP-1; IBP 1; IBP1; IBP-1; IGF BP25; IGFBA; IGF-binding protein 1; IGFBP; IGFBP 1; Igfbp1; Igfbp-1; IGF-BP25; Insulin like growth factor binding protein; insulin like growth factor binding protein 1; insulin-like growth factor binding protein 1; INSULIN-LIKE GROWTH FACTOR BINDING PROTEIN 1 PRECURSOR (IGFBP-1) (IBP-1) (IGF-BINDING PROTEIN 1); insulin-like growth factor-binding protein 1; Placental protein 12; PP 12; PP12 | |
| Human recombinant protein fragment corresponding to amino acids 97-312 of human CIAPIN1 produced in E.coli. | |
| 30 μL | |
| Primary | |
| Human | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction