missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IGFBP1 Monoclonal Antibody (OTI6C7), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA808733
This item is not returnable.
View return policy
Description
This gene is a member of the insulin-like growth factor binding protein (IGFBP) family and encodes a protein with an IGFBP domain and a thyroglobulin type-I domain. The protein binds both insulin-like growth factors (IGFs) I and II and circulates in the plasma. Binding of this protein prolongs the half-life of the IGFs and alters their interaction with cell surface receptors.Specifications
| IGFBP1 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| P08833 | |
| IGFBP1 | |
| Human recombit protein fragment corresponding to amino acids 26-259 of human IGFBP1 produced in E.coli. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Immunohistochemistry (Paraffin), Western Blot | |
| OTI6C7 | |
| Unconjugated | |
| IGFBP1 | |
| AFBP, IBP1, IGF-BP25, PP12, hIGFBP-1 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 3484 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction