missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IGFBP4 Rabbit, anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 30952-1-AP-20UL
This item is not returnable.
View return policy
Description
The IGFBP4 antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human IGFBP4. This antibody recognizes human antigen. The IGFBP4 antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
| IGFBP4 | |
| 400 μg/mL | |
| Rabbit | |
| Primary | |
| Human | |
| 150 kDa | |
| IgG |
| Western Blot, ELISA | |
| BP 4, HT29 IGFBP, IBP 4, IBP4, IGF binding protein 4, IGFBP 4, IGFBP4 | |
| 20 μL | |
| 3487 | |
| Store at -20°C. Stable for one year after shipment. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction