missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ IGFBP7 Polyclonal Antibody

Goat Polyclonal Antibody
Brand: Invitrogen™ PA547123
Denne vare kan ikke returneres.
Se returpolitik
Beskrivelse
In direct ELISAs, less than 1% cross-reactivity with recombinant human (rh) IGFBP-1, rhIGFBP-2, rhIGFBP-3, rhIGFBP-4, rhIGFBP-5, rhIGFBP-6, rhIGFBP-L1, rhIGFBP-rp10 and recombinant mouse IGFBP-7 is observed. Reconstitute at 0.2 mg/mL in sterile PBS.
This gene encodes a member of the insulin-like growth factor (IGF)-binding protein (IGFBP) family. IGFBPs bind IGFs with high affinity, and regulate IGF availability in body fluids and tissues and modulate IGF binding to its receptors. This protein binds IGF-I and IGF-II with relatively low affinity, and belongs to a subfamily of low-affinity IGFBPs. It also stimulates prostacyclin production and cell adhesion. Alternatively spliced transcript variants encoding different isoforms have been described for this gene, and one variant has been associated with retinal arterial macroaneurysm.
Tekniske data
| IGFBP7 | |
| Polyclonal | |
| Unconjugated | |
| IGFBP7 | |
| AGM; angiomodulin; FSTL2; IBP7; IBP-7; IGF-binding protein 7; IGFBP; IGFBP rP1; Igfbp7; IGFBP-7; IGFBP-7v; IGFBPRP1; IGFBP-rP1; Insulin like growth factor binding protein; insulin like growth factor binding protein 7; insulin-like growth factor binding protein 7; insulin-like growth factor-binding protein 7; Mac25; MAC25 protein; PGI2-stimulating factor; prostacyclin-stimulating factor; PSF; RAMSVPS; TAF; Tumor-derived adhesion factor | |
| Goat | |
| Antigen affinity chromatography | |
| RUO | |
| 3490 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Lyophilized |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.2 mg/mL | |
| PBS with 5% trehalose and No Preservative | |
| Q16270 | |
| IGFBP7 | |
| Mouse myeloma cell line NS0-derived recombinant human IGFBP-rp1 Arg98-Arg277. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
Ser du en mulighed for forbedring?Del en indholdskorrektion