missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IgG Antibody (B33/20), DyLight 680, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-47724FR
This item is not returnable.
View return policy
Description
IgG Monoclonal antibody specifically detects IgG in Human samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin), Immunofluorescence, CyTOF
Specifications
| IgG | |
| Monoclonal | |
| DyLight 680 | |
| Immunoglobulin G, ImmunoglobulinG | |
| Purified polyclonal human Ig Gamma Chain | |
| 0.1 mL | |
| Adaptive Immunity, Epitope Tags, Immunology | |
| 3500 | |
| Store at 4°C in the dark. | |
| IgG1 κ |
| Flow Cytometry, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunofluorescence | |
| B33/20 | |
| 50 mM Sodium Borate | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction