missing translation for 'onlineSavingsMsg'
Learn More

Armenian Hamster IgG, FITC, clone: eBio299Arm, Isotype Control, eBioscience™

Armenian Hamster Isotype Control

Brand:  Affymetrix eBioscience 11-4888-85

Additional Details : Weight : 0.25000kg

 View more versions of this product

Product Code. 15268229

  • € 232.00 / 500µg


Estimated Shipment
29-09-2020
Add to basket

Description

Description

Description: The eBio299Arm monoclonal antibody is useful as an isotype control immunoglobulin.Applications Reported: This eBio299Arm antibody has been reported for use in immunocytochemistry, immunohistochemistry, and flow cytometric analysis.Applications Tested: This eBio299Arm antibody has been tested by flow cytometric analysis of mouse splenocytes and normal human peripheral blood. Use isotype control at the same concentration as experimental antibody.Excitation: 488 nm; Emission: 520 nm; Laser: Blue Laser.Filtration: 0.2μm post-manufacturing filtered.
Specifications

Specifications

Armenian Hamster IgG
Control
0.5mg/mL
PBS with 0.1% gelatin and 0.09% sodium azide; pH 7.2
IgG
IgG
500μg
Store at 2-8°C. Do not freeze. Light-sensitive material.
Monoclonal
Control, Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry
eBio299Arm
FITC
Liquid
Armenian Hamster
Affinity chromatography
RUO
Isotype Control
Not Applicable
Videos
Documents

For Research Use Only.