Learn More
IgG Fc Goat anti-Rabbit, Polyclonal , Abnova™
Goat anti-rabbit IgG recognizes rabbit IgG F(c) fragment.
Brand:  Abnova PAB9399.50mg
Product Code. 16264670
  • € 997.00 / 50mg
Specifications
IgG Fc
Unconjugated
The optimal working dilution should be determined by the end user.
Goat
This product is an IgG fraction antibody purified from monospecific antiserum by a multi-step process which includes delipidation, salt fractionation and ion exchange chromatography followed by extensive dialysis against the buffer stated above.
RUO
No reaction was observed against rabbit IgG F(ab')2.
Store at 4°C on dry atmosphere. After reconstitution with deionized water store at −20°C or lower. Aliquot to avoid repeated freezing and thawing.
Polyclonal
Goat anti-rabbit IgG recognizes rabbit IgG F(c) fragment. This secondary antibody was purified using a multi-step process which includes delipidation, salt fractionation and ion exchange chromatography followed by extensive dialysis against the buffer stated above.
In 10mM sodium phosphate, 150mM NaCl, pH 7.2
Rabbit IgG F(c) fragment
50 mg
Secondary
Rabbit
Lyophilized