missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IgG Light Chain Antibody (IVA285-1), PE, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-62190PE
This item is not returnable.
View return policy
Description
IgG Light Chain Monoclonal specifically detects IgG Light Chain in Bovine, Sheep samples. It is validated for Flow Cytometry.
Specifications
| IgG Light Chain | |
| Monoclonal | |
| PE | |
| PBS with 0.05% Sodium Azide | |
| Fraction of Bovine immunoglobulin. | |
| 0.1 mL | |
| Bovine, Sheep | |
| IgG1 |
| Flow Cytometry | |
| IVA285-1 | |
| Flow Cytometry | |
| Mouse | |
| Protein A purified | |
| Primary | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction