missing translation for 'onlineSavingsMsg'
Learn More

Mouse IgG1 kappa Isotype Control (P3.6.2.8.1), PE-Cyanine7, eBioscience™, Invitrogen™

Mouse Isotype Control

Brand:  Affymetrix eBioscience 25-4714-80

89.33 EUR valid until 2024-03-29
Use promo code "21615" to get your promotional price.



Additional Details : Weight : 0.25000kg

 View more versions of this product

Product Code. 15330960

  • € 134.00 / 25µg
In Stock
to see stock.
Add to basket

Description

Description

Description: The monoclonal mouse IgG1 K immunoglobulin is useful as an isotype control. Applications Reported: PE-Cy7 Mouse IgG1, K Iso Cntrl has been reported for use in flow cytometric analysis. Applications Tested: This P3.6.2.8.1 antibody has been tested by flow cytometric analysis of normal human peripheral blood cells. This can be used at the same concentration as the experimental antibody. Light sensitivity: This tandem dye is sensitive photo-induced oxidation. Please protect this vial and stained samples from light. Fixation: Samples can be stored in IC Fixation Buffer (cat. 00-8222) (100 μL cell sample + 100 μL IC Fixation Buffer) or 1-step Fix/Lyse Solution (cat. 00-5333) for up to 3 days in the dark at 4°C with minimal impact on brightness and FRET efficiency/compensation. Some generalizations regarding fluorophore performance after fixation can be made, but clone specific performance should be determined empirically. Excitation: 488-561 nm; Emission: 775 nm; Laser: Blue Laser, Green Laser, Yellow-Green Laser. Filtration: 0.2 μm post-manufacturing filtered.

Mouse IgG1 kappa Control antibody specifically detects Mouse IgG1 kappa in Not Applicable samples. It is validated for Control, Flow Cytometry.
Specifications

Specifications

Mouse IgG1 kappa
P3.6.2.8.1
PE-Cyanine7
IgG1; Igh-4; Ighg1; Immunoglobulin G; Immunoglobulin G1; immunoglobulin heavy constant gamma 1 (G1m marker); ImmunoglobulinG; ImmunoglobulinG1; VH7183
Affinity chromatography
RUO
Not Applicable
Liquid
Control, Flow Cytometry
0.2 mg/mL
PBS with 0.09% sodium azide; pH 7.2
Mouse
25 μg
Isotype Control (Primary)
4° C, store in dark, DO NOT FREEZE!
IgG1 κ
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

For Research Use Only.