missing translation for 'onlineSavingsMsg'
Learn More

Rat IgG1 kappa Isotype Control (eBRG1), eFluor™ 450, eBioscience™, Invitrogen™

Rat Isotype Control

Brand:  Affymetrix eBioscience 48-4301-82

140.00 EUR valid until 2024-03-29
Use promo code "21615" to get your promotional price.



Additional Details : Weight : 0.25000kg

 View more versions of this product

Product Code. 15331770

  • € 210.00 / 100µg
In Stock
to see stock.
Add to basket

Description

Description

Description: The monoclonal rat IgG1, kappa is useful as an isotype control immunoglobulin. Applications Reported: Rat IgG1 Isotype Control eFluor® 450 has been reported for use in flow cytometric analysis. Applications Tested: eFluor® 450 Rat IgG1 Isotype Control has been tested by flow cytometric analysis of mouse splenocytes. Use isotype control at the same concentration as experimental antibody. eFluor® 450 is an alternative to Pacific Blue®. eFluor® 450 emits at 445 nm and is excited with the Violet laser (405 nm). Please make sure that your instrument is capable of detecting this fluorochome. Excitation: 405 nm; Emission: 445 nm; Laser: Violet Laser. Filtration: 0.2 μm post-manufacturing filtered.

Rat IgG1 kappa Control antibody specifically detects Rat IgG1 kappa in Not Applicable samples. It is validated for Control, Flow Cytometry.
Specifications

Specifications

Rat IgG1 kappa
eBRG1
eFluor 450
Immunoglobulin G; Immunoglobulin G1; ImmunoglobulinG; ImmunoglobulinG1
Affinity chromatography
RUO
Not Applicable
Liquid
Control, Flow Cytometry
0.2 mg/mL
PBS with 0.09% sodium azide; pH 7.2
Rat
100 μg
Isotype Control (Primary)
4° C, store in dark, DO NOT FREEZE!
IgG1 κ
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

For Research Use Only.