Learn More
IgG2b Mouse, APC, Clone: MPC-11, Isotype Control, STEMCELL Technologies
Mouse Monoclonal Antibody
Brand:  Stemcell Technologies 60072AZ.1
Encompass_Preferred
 View more versions of this product
Product Code. 17128861
  • € 68.85 / 25µg
Description
  • The MPC-11 antibody (IgG2b, kappa) is suitable for use as an isotype-matched control antibody in several applications to estimate the degree of non-specific binding by an antigen-specific antibody
  • Ideally, the isotype control should have the same subclass of heavy chain (IgA, IgD, IgE, IgG, or IgM) and light chain (kappa or lambda) as the specific antibody being used
  • If a conjugated antibody is used, an isotype control conjugated to the same molecule (e.g. fluorochrome) should be chosen
  • The use of an appropriate isotype control helps confirm the specificity of the antigen-specific antibody, and indicates non-specific binding that may result from binding to Fc receptors or other cell components
  • This antibody clone has been verified for use as an isotype control antibody for assessing non-specific binding to cells in applications such as flow cytometry and immunofluorescence microscopy applications (surface and intracellular staining)
Specifications
Specifications
IgG2b
Monoclonal
APC
25 μg
IgG2b κ
Immunohistochemistry, Western Blot, ELISA, Flow Cytometry, Immunocytochemistry
MPC-11
Mouse
Isotype Control (Primary)
Other