Learn More
IgG2b Mouse, APC, Clone: MPC-11, Isotype Control, STEMCELL Technologies
Mouse Monoclonal Antibody
Brand: Stemcell Technologies 60072AZ.1
This item is not returnable.
View return policy
Description
- The MPC-11 antibody (IgG2b, kappa) is suitable for use as an isotype-matched control antibody in several applications to estimate the degree of non-specific binding by an antigen-specific antibody
- Ideally, the isotype control should have the same subclass of heavy chain (IgA, IgD, IgE, IgG, or IgM) and light chain (kappa or lambda) as the specific antibody being used
- If a conjugated antibody is used, an isotype control conjugated to the same molecule (e.g. fluorochrome) should be chosen
- The use of an appropriate isotype control helps confirm the specificity of the antigen-specific antibody, and indicates non-specific binding that may result from binding to Fc receptors or other cell components
- This antibody clone has been verified for use as an isotype control antibody for assessing non-specific binding to cells in applications such as flow cytometry and immunofluorescence microscopy applications (surface and intracellular staining)
Specifications
| IgG2b | |
| Monoclonal | |
| APC | |
| 25 μg | |
| IgG2b κ |
| Immunohistochemistry, Western Blot, ELISA, Flow Cytometry, Immunocytochemistry | |
| MPC-11 | |
| Mouse | |
| Isotype Control (Primary) | |
| Other |