Learn More
IgG2b Mouse, APC, Clone: MPC-11, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech APC-65128-100UG
Description
The IgG2b Isotype Control antibody from Proteintech is a mouse monoclonal antibody generated with n/a of mouse IgG2b Isotype Control. This antibody recognizes antigen. The IgG2b Isotype Control antibody has been validated for the following applications: FC analysis. This antibody is apc.
Specifications
| IgG2b | |
| Monoclonal | |
| 0.2 mg/mL | |
| Mouse | |
| Primary | |
| IgG2b |
| Flow Cytometry | |
| MPC-11 | |
| APC | |
| 100 μg | |
| 16016 |