Learn More
IgG2b Mouse, APC, Clone: MPC-11, Proteintech
Mouse Monoclonal Antibody
Brand:  Proteintech APC-65128-100UG
Product Code. 16850506
  • € 48.00 / 100µg
Description
The IgG2b Isotype Control antibody from Proteintech is a mouse monoclonal antibody generated with n/a of mouse IgG2b Isotype Control. This antibody recognizes antigen. The IgG2b Isotype Control antibody has been validated for the following applications: FC analysis. This antibody is apc.
Specifications
Specifications
IgG2b
Monoclonal
0.2 mg/mL
Mouse
Primary
IgG2b
Flow Cytometry
MPC-11
APC
100 μg
16016