missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IgG4 Fc Antibody (RM217) - Azide and BSA Free, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-25978
This item is not returnable.
View return policy
Description
IgG4 Fc Monoclonal antibody specifically detects IgG4 Fc in Human samples. It is validated for ELISA, ELISA, ELISA
Specifications
| IgG4 Fc | |
| Monoclonal | |
| Unconjugated | |
| PBS | |
| Peptide corresponding to the Fc region of Human IgG4 | |
| 100 μg | |
| Primary | |
| Store at -20C. Avoid freeze-thaw cycles. | |
| IgG |
| ELISA | |
| RM217 | |
| ELISA, Sandwich ELISA Capture : 50 - 200ng/well, Sandwich ELISA Detection : 0.05 - 0.2ug/ml | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction