missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IgG4 Mouse anti-Human, Clone: OTI3G2, TrueMAB™, Origene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA814478S
This item is not returnable.
View return policy
Description
IgG4 Monoclonal antibody specifically detects IgG4 in Human samples. It is validated for ELISASpecifications
| IgG4 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| P01861 | |
| Mouse | |
| Protein A/G | |
| RUO | |
| 3503 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA | |
| OTI3G2 | |
| Unconjugated | |
| IGHG4 | |
| IGHG4; Immunoglobulin G; immunoglobulin heavy constant gamma 4 (G4m marker); ImmunoglobulinG; ImmunoglobulinG4 | |
| Native Human IgG4 protein | |
| 30 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction