Learn More
IgG4 Mouse, Clone: IGHG4/1345, enQuireBio™
Mouse Monoclonal Antibody
Brand: EnquireBio 3503-MSM1-P0
This item is not returnable.
View return policy
Specifications
| IgG4 | |
| Cytoplasmic | |
| IGHG4/1345 | |
| Unconjugated | |
| IGHG4 | |
| Recombinant human full-length IGHG4 | |
| Tonsil | |
| 20 μg | |
| Primary | |
| Short term: Store at 2-4°C. Long term, aliquot and store at -20°C. |
| Immunohistochemistry | |
| Monoclonal | |
| 200μg/mL | |
| 10mM PBS with 0.05% BSA and 0.05% Azide | |
| Mouse | |
| 150kDa | |
| Protein A/G Purified | |
| RUO | |
| 3503 | |
| IgG1 |