Learn More
IGHD Mouse anti-Human, CoraLite 488, Clone: 1D1B12, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech CL488-67538-100UL
Additional Details : Weight : 0.00010kg
Description
IgD is the major antigen receptor isotype on the surface of most peripheral B-cells. The function of IgD is to signal the B cells to be activated. The relative molecular mass and half-life of secreted IgD is 185 kDa and 2.8 days, respectively. Secreted IgD is produced as a monomeric antibody with two heavy chains of the delta (δ) class, and two Ig light chains. This antibody detects the heavy chain (55-70 kDa) of IgD and secreted IgD (185 kDa) .
Specifications
IGHD | |
Monoclonal | |
1000 μg/mL | |
Mouse | |
Primary | |
Human | |
IgG2a |
Immunofluorescence | |
1D1B12 | |
CoraLite 488 | |
100 μL | |
3495 | |
Liquid |