Learn More
IGJ Mouse anti-Human, Clone: OTI3E12, lyophilized, TrueMAB™
Mouse Monoclonal Antibody
Brand: Origene Technologies CF504172
Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
J chain is a small glycopeptide and is structurally unrelated to heavy or light chains, but is synthesized by all plasma cells that secrete polymeric immunoglobulins. J chain is linked to IgA and IgM by disulfide bonds, however, it has been detected in IgG- and IgD-containing cells. J chains are present in a large proportion of the immunoglobulin-positive cells in the germinal centers of the tonsil and lymph node. B cells secrete J chain at an early stage of differentiation with the expression persisting in those cells destined to produce IgA or IgM.Specifications
| IGJ | |
| Monoclonal | |
| Unconjugated | |
| JCHAIN | |
| IGCJ, IGJ, JCH | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 3512 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Lyophilized |
| Flow Cytometry, Western Blot | |
| OTI3E12 | |
| PBS with 8% trehalose and no preservative; pH 7.3 | |
| P01591 | |
| JCHAIN | |
| Full length human recombit protein of human IGJ produced in HEK293T cell. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.