missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IgM Antibody (DA4-4 (same as SA-DA4 or HB57)), DyLight 594, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-34601DL594
This item is not returnable.
View return policy
Description
IgM Monoclonal antibody specifically detects IgM in Human samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin), CyTOF
Specifications
| IgM | |
| Monoclonal | |
| DyLight 594 | |
| AGM1, DKFZp686I15196, DKFZp686I15212, FLJ00385, immunoglobulin heavy constant mu, MGC104996, MGC52291, MU, VH | |
| Heavy chain of human IgM | |
| 0.1 mL | |
| B Cell Development and Differentiation Markers, Immunology | |
| 3507 | |
| Store at 4°C in the dark. | |
| IgG1 κ |
| Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry (Paraffin), CyTOF | |
| DA4-4 (same as SA-DA4 or HB57) | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction