missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IgM Antibody (DA4-4 (same as SA-DA4 or HB57)), PE/Cy7, Novus Biologicals™
Mouse Monoclonal Antibody
Marca: Novus Biologicals NBP2-34601PECY7
Les retours ne sont pas autorisés pour ce produit.
Consulta la politica di reso
Descrizione
IgM Monoclonal specifically detects IgM in Human samples. It is validated for Flow Cytometry.
Specifica
| IgM | |
| Monoclonal | |
| PE-Cyanine7 | |
| PBS with 0.05% Sodium Azide | |
| IGHM | |
| Heavy chain of human IgM | |
| 0.1 mL | |
| Primary | |
| Recognizes a protein of 75kDa, identified as mu heavy chain of human immunoglobulins. It does not cross-react with alpha (IgA), gamma (IgG), epsilon (IgE), or delta (IgD), heavy chains, T-cells, monocytes, granulocytes, or erythrocytes. This monoclonal antibody is useful in the identification of leukemias, plasmacytomas, and certain non-Hodgkins lymphomas. The most common feature of these malignancies is the restricted expression of a single heavy chain class. Demonstration of clonality in lymphoid infiltrates indicates that the infiltrate is clonal and therefore malignant. | |
| Store at 4°C in the dark. Do not freeze. |
| Flow Cytometry | |
| DA4-4 (same as SA-DA4 or HB57) | |
| Flow Cytometry | |
| AGM1, DKFZp686I15196, DKFZp686I15212, FLJ00385, immunoglobulin heavy constant mu, MGC104996, MGC52291, MU, VH | |
| Mouse | |
| Protein A or G purified | |
| B Cell Development and Differentiation Markers, Immunology | |
| 3507.0 | |
| Human | |
| IgG1 κ |
Correzione del contenuto del prodotto
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
Titolo del prodotto
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto